missing translation for 'onlineSavingsMsg'
Learn More

Piperacillin, 95%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66143.03


Product Code. 15457389

  • ONLINE EXCLUSIVE PRICE
    169.00€
    152.65€ / 1g
    Save 16.35€
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

61477-96-1
1g
MFCD00865043
WCMIIGXFCMNQDS-RAMASXNTSA-M
sodium (2R,5S,6S)-6-[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
43672
517.55
Piperacillin
White
C23H26N5NaO7S
Freely soluble in methanol; Only sparingly soluble in aqueous solution at 0;119mg/mL
[Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@@H]2[C@@H]3SC(C)(C)[C@H](N3C2=O)C([O-])=O)C2=CC=CC=C2)C(=O)C1=O
539.54
CHEBI:8232
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Piperacillin

Signal Word
  • Danger

Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contain: Piperacillin

SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.