Learn More
Cefotiam dihydrochloride, Thermo Scientific™
A broad spectrum antibiotic
Brand:  Thermo Scientific Alfa Aesar J66529.MC
Product Code. 15467929
  • 142.00€ / 100mg
 Restricted Item
Estimated Shipment: 01-10-2026
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
66309-69-1
100mg
C18H27Cl2N9O5S3
94°C
KHUCCKUOJMXYKH-BUWDDJPCSA-N
(6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
70590037
Cefotiam dihydrochloride
White
>110°C (230°F)
MFCD09878605
Soluble in water
CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
616.552
562.09
Product Suggestions
Product Suggestions
Videos
SDS Documents