Learn More
CGS 15943, 99+%, Thermo Scientific™
Potent adenosine receptor antagonist
Brand: Thermo Scientific Alfa Aesar J61313.MA
| Quantity | 10mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 104615-18-1 | |
| MFCD01529897 | |
| 9-chloro-2-2-furyl-1,2,4 triazolo 1,5-c quinazolin-5-imine, 9-chloro-2-furan-2-yl-1,2,4 triazolo 1,5-c quinazolin-5-amine, chembl16687, 9-chloro-2-2-furyl 1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine,9-chloro-2-2-furanyl, 9-chloro-2-2-furanyl-1,2,4 triazolo 1,5-c quinazolin-5-amine, 1,2,4 triazolo 1,5-c quinazolin-5-amine, 9-chloro-2-2-furanyl, acmc-20ddlj | |
| NC1=NC2=CC=C(Cl)C=C2C2=NC(=NN12)C1=CC=CO1 | |
| 285.69 | |
| 285.69 | |
| CGS 15943 |
| C13H8ClN5O | |
| 10mg | |
| MSJODEOZODDVGW-UHFFFAOYSA-N | |
| 9-chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]quinazolin-5-amine | |
| 2690 | |
| ≥99% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.