missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 1070722
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
Description
Potent glycogen synthase kinase-3 (GSK-3) inhibitor (Ki = 0.6 nM for GSK-3α and GSK-3β). Displays high selectivity (> 50-fold) for GSK-3 over a panel of other kinases tested, including CDK family members. Decreases phosphorylation of microtubule-associated protein Tau in vitro; protects rat primary cortical neurons against β amyloid and glutamate challenge. Brain penetrant.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 362.31 |
| Purity | >99% |
| Molecular Weight (g/mol) | 362.312 |
| Chemical Name or Material | A 1070722 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction