missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 1899

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
A 1899 is a potent K2P3.1 (TASK-1) and K2P9.1 (TASK-3) channel blocker (IC50 values are 7 nM and 70 nM for human TASK-1 and TASK-3 expressed in CHO cells, respectively). Displays >12.5-fold selectivity for TASK-1 and TASK-3 over other K+ channels (IC50 = 0.9 μM for K2P18.1/TRESK and >2 μM for all other channels tested). Binds within open channel pore.
Specifications
Specifications
| Chemical Name or Material | N-[(2,4-Difluorophenyl)methyl]-2'-[[[2-(4-methoxyphenyl)acetyl]amino]methyl][1,1'-biphenyl]-2-carboxamide |
| CAS | 498577-46-1 |
| Quantity | 5 mg |
| Molecular Formula | C30H26F2N2O3 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction