missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ A 1899
GREENER_CHOICE

Product Code. p-7180145
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18726426

Brand: Tocris Bioscience™ 6972/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent K2P3.1 (TASK-1) and K2P9.1 (TASK-3) blocker

A 1899 is a potent K2P3.1 (TASK-1) and K2P9.1 (TASK-3) channel blocker (IC50 values are 7 nM and 70 nM for human TASK-1 and TASK-3 expressed in CHO cells, respectively). Displays >12.5-fold selectivity for TASK-1 and TASK-3 over other K+ channels (IC50 = 0.9 μM for K2P18.1/TRESK and >2 μM for all other channels tested). Binds within open channel pore.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(2,4-Difluorophenyl)methyl]-2'-[[[2-(4-methoxyphenyl)acetyl]amino]methyl][1,1'-biphenyl]-2-carboxamide
CAS 498577-46-1
Quantity 5 mg
Molecular Formula C30H26F2N2O3
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.