missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 71915

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
A 71915 is a highly potent and competitive natriuretic peptide receptor A (NPRA) antagonist (pKi = 9.18). Reduces brain natriuretic peptide ( BNP, Cat. No. 3522 )-induced scratching in mice.
Specifications
Specifications
| CAS | 132956-87-7 |
| Content And Storage | Store at -20°C |
| Description | Natriuretic peptide receptor A (NPRA) antagonist |
| Formulation | C69H116N26O15S2 |
| Molecular Weight (g/mol) | 1613.95 |
| Quantity | 1 mg |
| Sequence | RCXGGRIDRIXRC (Modifications: X-3 = Cha, X-11= D-Tic, Cys-13 = C-terminal amide, Disulfide bridge between Cys-2 - Cys-13) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction