Learn More
Tocris Bioscience™ A 77-01
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
A 77-01 is a potent TGF-βRI (ALK-5) inhibitor (IC50 = 34 nM). Inhibits TGF-β transcriptional activation. Inhibits TGF-β-induced proliferation of lung epithelial cells and SMAD phosphorylation. Inhibits TGF-β-induced epithelial-to-mesenchymal transition (EMT). Also inhibitor of p38 MAPK. Likely active metabolite of A 83-01 (Cat. No. 2939).
Specifications
Specifications
| Chemical Name or Material | 4-[3-(6-Methyl-2-pyridinyl)-1H-pyrazol-4-yl]quinoline |
| CAS | 607737-87-1 |
| Quantity | 25 mg |
| Molecular Formula | C18H14N4 |
| Formula Weight | 286.33 |
| Purity | 98% |