missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies A 83-01
Description
- A 83-01 is a potent inhibitor of activin receptor-like kinase (ALK) including ALK5 (type I transforming growth factor-β receptor), ALK4 (type IB activin receptor), and ALK7 (type I NODAL receptor) with IC50 = 12, 45, and 7.5 nM, respectively (Tojo et al.)
- Facilitates the conversion of epiblast stem cells to the naїve pluripotent state (Zhou et al.; Li et al.)
- In combination with PD0325901, enables OCT4-mediated reprogramming (Zhu et al.)
- Inhibits mesoderm and endoderm specification (Zhang et al.)
- Disrupts epithelial to mesenchymal transition through inhibition of the SMAD signaling pathway (Tojo et al.)
Specifications
Specifications
| CAS | 909910-43-6 |
| Quantity | 5 mg |
| Molecular Formula | C25H19N5S |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?