missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ A 971432

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A 971432 is a potent and selective S1P5 agonist (EC50 values are 4.1 and 5.7 nM in cAMP and GTPγS assays, respectively). Exhibits 60-fold selectivity over S1P1, and >1,600-fold selectivity over S1P2-4, and a panel of 129 kinases and 79 receptors. Improves blood-brain barrier integrity in vitro and reverses age-related cognitive decline in vivo. Orally available.
Specifications
Specifications
| Chemical Name or Material | 1-[[4-[(3,4-Dichlorophenyl)methoxy]phenyl]methyl]-3-azetidinecarboxylic acid |
| CAS | 1240308-45-5 |
| Quantity | 10 mg |
| Target | Sphingosine-1-phosphate Receptor Agonists |
| Molecular Formula | C18H17Cl2NO3 |
| Formula Weight | 366.24 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction