Learn More
Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers
Specially designed for comfort, safety and strength. Kimberly-Clark™ KLEENGUARD™ A50 Breathable Splash and Particle Protection Trousers is designed for comfort, safety and general protection against larger particles, radioactive dust and chemical splashes.
Brand: Kimberly-Clark™ 99520
Additional Details : Weight : 0.01000kg
Description
- Approved as Complex Design (Category 3) equipment - offering protection to the levels specified for Type 5 and 6 by CEN
- Single use and breathable protective trousers designed for maximum comfort and safety, providing anti-static and general protection against water-based or chemical splashes, liquids, and dust particles
- Durable SMS fabric with additional treatment for increased protection
- Suitable for use as protection against solid particles and particulate radioactive contamination - EN1073-2
- Silicone-free, ultra-low lint and treated to be antistatic - to EN 1149-5
- Ideal for protecting against powders, larger fibres and protection against a wide range of chemicals - reducing contamination in critical areas
- Suitable for use in pharmaceutical industries, manufacturing, utilities, electronics, agriculture and paint spraying areas
Specifications
White | |
While working in chemical and pharmaceutical industries. | |
Double-layer fabric | |
0.01 kg |
X-Large | |
Larger particles, radioactive dust and chemical splashes | |
Trousers |