Learn More
Testo™ 926 TE, Type T Temperature Meter
Ideal digital temperature meter for checking core temperature of food in commercial kitchens, catering, and other industrial environments. Testo™ 926 TE, Type T Temperature Meter is quick, reliable, and discreet.
Brand: Testo™ 0560 9261
Additional Details : Weight : 0.17000kg
Description
- Connectivity for an attachable/wireless sensor
- Wide range of optional sensors to suit your individual needs
- User-defined thresholds / audible alarm alerts when threshold exceeded
- Certified according to HACCP and EN 13485 / ideal for use with food
Specifications
182 x 64 x 40 mm | |
-50°C | |
64 mm |
0.1°C (−50 - 199.9°C) | |
±0.3°C (−20 - 70°C) |