missing translation for 'onlineSavingsMsg'
Learn More

Ac-Tyr-Val-Ala-Asp-7-Amino-4-TriFluoromethylcoumarin, MP BIOMEDICALS

Product Code. 11419885 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 11419885

Brand: MP Biomedicals 03AFC12010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
  • Ac-Tyr-Val-Ala-Asp-7-Amino-4-Trifluoromethylcoumarin.
  • Short peptide.
  • Caspase-1 substrate.
  • Enzyme ativity detection.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
TRUSTED_SUSTAINABILITY

Specifications

Chromagen Type Fluorogenic Chromagen
Detection Method Fluorescence
Form Lyophilized
For Use With (Application) Study Caspase
Quantity 10 mg
Substrate Fluorogenic Substrate
Substrate Properties Peptide-Based Substrate
Substrate Type Caspase Substrate
Target Enzyme Proteases
Product Type Activators, Inhibitors and Substrates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.