Learn More
BINDER™ Left Access Port (10 mm)
For Use With: BINDER units series FED
Brand: BINDER™ 8012-1278
Additional Details : Weight : 1.90000kg
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 10 mm, left | |
Access Port | |
Silicone Plug |
BINDER Chambers Series FED | |
Left | |
10 mm |