Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: BINDER units Series M
Brand: BINDER™ 8012-1363
Additional Details : Weight : 0.00001kg
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 30 mm, left | |
Access Port | |
Silicone Plug |
BINDER Chambers Series M | |
Left | |
30 mm |