Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: All MKF Series models
Marque:  BINDER™ 8012-1323
Informations supplémentaires : Weight : 0.30000kg
Code produit. 15902972
  • 505.00€ / Each
Estimated Shipment: 14-06-2024
to see stock.
Ajouter au panier
Description
Description
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Spécification
Spécification
Access port with silicone plug - 30 mm, left
Access Port
30 mm
BINDER Chambers Series MKF
Silicone plug