Learn More
BINDER™ Left Access Port (10 mm)
For Use With: FDL 115
Brand:  BINDER™ 8012-1277
Additional Details : Weight : 0.28000kg
Product Code. 16404159
  • 386.00€ / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Specifications
Access port with silicone plug - 10 mm, left
Access Port
10 mm
BINDER Chambers FDL 115
Silicone plug