Learn More
BINDER™ Left Access Port (10 mm)
For Use With: FP 53, FP 115, FP 240, FP 400, FP 720
Brand: BINDER™ 8012-1279
Additional Details : Weight : 0.28000kg
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 10 mm, left | |
Access Port | |
10 mm |
BINDER Chambers Series FP | |
Silicone plug |