missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Left Access Port (10 to 30 mm)

For Use With: M 53, M 115, M 240, M 400, M 720

Brand:  BINDER™ 8012-1280

Additional Details : Weight : 0.28000kg

Product Code. 16424159

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

  • For use on left
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications

Specifications

Access port with silicone plug - 10 mm, left
Access Port
10 mm
BINDER Chambers Series M
Silicone plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions