Learn More
BINDER™ Left Access Port (10 to 30 mm)
For Use With: FD 23
Brand: BINDER™ 8012-1359
Additional Details : Weight : 0.28000kg
Description
- For use on left
- Includes silicone plug
- For use with a wide range of BINDER™ incubators and chambers
Specifications
Access port with silicone plug - 30 mm, left | |
Access Port | |
30 mm |
BINDER Chambers FD 23 | |
Silicone plug |