Learn More
Epredia™ Accessories for all Richard-Allan Scientific™ Cryostat Models, Flat Cryostat Brush, 10mm
Shop All Epredia Products
Click to view available options
Product Type:
Cryo-molds, 10mm
Cryo-molds, 15mm
Cryo-molds, 22mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Cryo-molds, 10mm |
| For Use With (Equipment) | Richard-Allan Scientific Cryostats |