Learn More
Thermo Scientific™ Accessories for Bottle/Tube Roller
Product Code. 15248914 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
O-ring for 15 mL tube
O-ring for 50 mL tube
Remote Control
Roller
Stacking Bar
Unit Size:
Each
Pack of 2
Pack of 20
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
15248914 O-ring for 50 mL tube Pack of 20
15238914 Roller Pack of 2
15258914 O-ring for 15 mL tube Pack of 20
15268914 Remote Control Each
15278914 Stacking Bar Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15248914 Supplier Thermo Scientific™ Supplier No. 88881011
Only null left
Add to basket
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Use these accessories to optimize the performance of your Thermo Scientific™ Bottle/Tube Roller.
Use Thermo Scientific™ Accessories for Bottle/Tube Roller to optimize flexibility and performance.
TRUSTED_SUSTAINABILITY
Tekniske data
Tube Size 50 mL
Product Type O-ring for 50 mL tube
Dimensions (L x W) 23.5 x 3.55 mm (0.93 x 0.14 in.)
Description O-ring for 50mL tube (pack of 20) 23.5 x 3.55mm 0.93 x 0.14 in.
For Use With (Equipment) Bottle/Tube Roller
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.