Learn More
Sartorius Accessories for Combisart™ Systems: Filtrate Flasks
Product Code. 10623941 Shop All Sartorius Products
Click to view available options
Capacity (Metric):
1000mL
2000mL
5000mL
Unit Size:
Each
Product Code. 10623941
Brand: Sartorius 16672
Only null left
Add to basket
Add to basket
Filtrate flasks for use with Sartorius Combisart Systems
  • Suction flask for individual filter holders
  • Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
  • Plastic hose nipple acc. to DIN 12476
  • Stopper/tube
  • Sterilizable
TRUSTED_SUSTAINABILITY
Specifications
Product Type Suction Flask
Display Type None
Capacity (Metric) 2000mL
Color Translucent
Material Borosilicate Glass
For Use With (Application) For collecting the filtrate
For Use With (Equipment) Individual vacuum filter holder
Includes 9 mm Plastic Safety Hose Nipple