Learn More
Sartorius Accessories for Combisart™ Systems: Filtrate Flasks
Shop All Sartorius Products
Click to view available options
Capacity (Metric):
1000mL
2000mL
5000mL
Unit Size:
Each
Description
Filtrate flasks for use with Sartorius Combisart Systems
- Suction flask for individual filter holders
- Vacuum-resistant borosilicate glass, 3.3 (Duran 50 glass)
- Plastic hose nipple acc. to DIN 12476
- Stopper/tube
- Sterilizable
Specifications
Specifications
| Product Type | Suction Flask |
| Display Type | None |
| Capacity (Metric) | 5000mL |
| Color | Translucent |
| Material | Borosilicate Glass |
| For Use With (Application) | For collecting the filtrate |
| For Use With (Equipment) | Individual vacuum filter holder |
| Includes | Plastic Hose Nipple |