missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Dessicator Shelves for Fiber Glass Desiccator

Two aluminum shelves for use with Labconco Vacuum Desiccator.

Brand:  Labconco™ 5532000

Additional Details : Weight : 0.80000kg

Product Code. 11781379

  • 117.00€ / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket

Description

Description

  • Two aluminum shelves, each with twelve 7/8 in. (22 mm holes) to support small containers such as crucibles
Specifications

Specifications

Desiccator Shelf
Labconco Fiber Glass Desiccator
1.8 kg
Aluminum
2 shelves
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions