Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in a wide range of applications and in conjunction with our various lines of filtration devices.
18.50€ - 3512.00€
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10182541
|
Thermo Scientific™
710300-0001 |
O-Rings and Gaskets |
36.40€
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
10090352
|
Thermo Scientific™
710300-0004 |
O-Rings |
37.30€
Each |
In Stock
Log in to see stock. |
|||||
10693252
|
Thermo Scientific™
710300-0005 |
Vacuum Caps and Tubing Adapter |
18.50€
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10132641
|
Thermo Scientific™
710300-0006 |
Vacuum Tubing Adapters |
20.80€
Pack of 2 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||||
10471022
|
Thermo Scientific™
710320-0045 |
Bottle top gasket, outer diameter 424mm (1.67 inch), 5 pieces |
57.90€
Pack of 5 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
10440143
|
Thermo Scientific™
DS0315-0047 |
250mL |
223.00€
Each |
In Stock
Log in to see stock. |
|||||
10634212
|
Thermo Scientific™
DS0390-0070 |
Overal height w/adapter: 114mm; Bell I.D.: 70mm |
44.70€
Each |
In Stock
Log in to see stock. |
|||||
10673062
|
Thermo Scientific™
DS0395-0708 |
Vacuum Gasket O.D. x H x Thickness: 1.56 to 1.09 x 0.18 x 0.109 in. |
74.80€
Pack of 6 |
In Stock
Log in to see stock. |
|||||
10315751
|
Thermo Scientific™
DS0345-0001 |
Vacuum Filter Funnel Manifold |
1802.00€
Each |
Estimated Shipment: 19-11-2024
Log in to see stock. |
|||||
10243971
|
Thermo Scientific™
DS0397-0010 |
Filter Funnel Adapter |
52.20€
Pack of 25 |
In Stock
Log in to see stock. |
|||||
10193871
|
Thermo Scientific™
DS0405-0050 |
For all 50, 75 and 90mm MF75 filter units and bottle top filters |
3512.00€
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
|||||
10010431
|
Thermo Scientific™
DS0399-0001 |
Stainless-Steel Forceps, bent tip |
48.30€
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
10112161
|
Thermo Scientific™
DS0399-0002 |
Stainless-Steel Forceps, straight tip |
43.20€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
12085097
|
Thermo Scientific™
DS0396-0080 |
Rubber Stopper, number 8 with 5/8in hole |
42.30€
Pack of 3 |
In Stock
Log in to see stock. |
|||||
10052201
|
Thermo Scientific™
710315-0003 |
Cover with 3 TPE Caps |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
12746697
|
Thermo Scientific™
710345-0010 |
Filter Manifold Stopcock |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
10566551
|
Thermo Scientific™
710300-0003 |
Sterilization Plates and O-Rings |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.