Learn More
Merck Accessories for Samplicity™ Filtration Systems
Product Code. p-7232203 Shop All Merck Products
Click to view available options
Type:
Replacement Lid
Tube Set Assembly
Material:
Silicone
Thermoplastic Elastomer
Unit Size:
Each
Product Code. 16641744
Brand: Merck SAMP2LID
Only null left
Add to basket
Add to basket
To be used with the Samplicity™ G2 Filtration System.
Powered by vacuum, the system takes minimal manual effort to filter 1 to 8 samples into standard HPLC vials in seconds, with higher recovery than syringe filters provide.
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Description Nonsterile lid with doors for attachment of Millex filters/funnels to HPLC tubes
For Use With (Equipment) Samplicity G2 Filtration System with Millex Filters
Material Thermoplastic Elastomer
For Use With (Application) Analytical sample prep; High Performance Liquid Chromatography (HPLC)
Housing Material PVDF
Quantity 1 Ea.
Sufficient For Up to 8 samples
Type Replacement Lid