Learn More
WTW™ Accessory Box
GREENER_CHOICE
Product Code. 10171721
Change view
Click to view available options
For Use With (Equipment):
Ammonia Electrode NH 500/2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10171721 Ammonia Electrode NH 500/2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10171721 Supplier WTW™ Supplier No. 180100
Specifications
For Use With (Equipment) Ammonia Electrode NH 500/2
Type Accessory Box
Includes 3 Exchange Heads and 50mL Electrolyte Solution
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.