Learn More
Grant Instruments™ Cooling Coil
Provides a source of constant cooling to enable bath to be operated at or below ambient, down to 0°C. Grant Instruments™ Cooling Coil is for use with OLS Aqua Pro Orbital and Linear Shaking Water Bath.
Brand:  Grant Instruments™ CC26R
Additional Details : Weight : 17.00000kg
Product Code. 15315872
  • 7526.00€ / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
Provides a source of constant cooling to enable bath to be operated at or below ambient, down to 0°C. Grant Instruments™ Cooling Coil is for use with OLS Aqua Pro Orbital and Linear Shaking Water Bath.
  • For use with OLS Aqua Pro
  • Temperature range: Ambient to 0°C
Specifications
Specifications
Cooling coil accessory for OLS Aqua Pro linear shaking bath
OLS Aqua Pro
0°C
Accessory Cooling Coil