Learn More
Grant Instruments™ Cooling Coil
Provides a source of constant cooling to enable bath to be operated at or below ambient, down to 0°C. Grant Instruments™ Cooling Coil is for use with OLS Aqua Pro Orbital and Linear Shaking Water Bath.
Brand: Grant Instruments™ CC26R
Description
Provides a source of constant cooling to enable bath to be operated at or below ambient, down to 0°C. Grant Instruments™ Cooling Coil is for use with OLS Aqua Pro Orbital and Linear Shaking Water Bath.
- For use with OLS Aqua Pro
- Temperature range: Ambient to 0°C
Specifications
Cooling coil accessory for OLS Aqua Pro linear shaking bath | |
OLS Aqua Pro | |
0°C | |
Accessory Cooling Coil |