Learn More
Thermo Scientific™ Acclaim™ PepMap™ C18 Peptide Analysis HPLC Column, 5 μm, 0.2 mm x 10 mm
Product Code. 11322253 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11322253 10 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11322253 Supplier Thermo Scientific™ Supplier No. 164212
Only null left
Add to basket
Add to basket
Column Format: Pre-Column Traps; Carbon Load: 15%; Pore Size: 100 Å; Column Phase: Reversed Phase
Offer high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ Acclaim™ 5μm PepMap™ 100 Nano-Trap Columns.
  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows hghest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Typically applied for the desalting of peptides before LC separation with MS detection
TRUSTED_SUSTAINABILITY
Specifications
Column Format Pre-Column Traps
Diameter (Metric) 0.2 mm
For Use With LC/MS, ESI-MS, MALDI-MS
Length (Metric) 10 mm
Max. Pressure 14500 psi (1000 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Surface Area 450 m²/g
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Silica
Carbon Load 15%
Column Type Peptide Analysis
Phase Reversed Phase
Material Fused Silica