Description
For accu-jet™ S:
- Part No. 26539: Plastic wall support
- Part No. 26606: Universal AC Adapter, 100-240V / 50-60Hz
- Part No. 26547: Snape tape
For accu-jet™ pro:
- Part No. 26540: Plastic wall support
For accu-jet™ S and accu-jet™ S:
- Part No: 26630: NiMH Battery
- Part No. 26508: Silicone Adapter, with non-return valve, 54 mm
Specifications
Specifications
| Battery Type | NiMH |
| For Use With | accu-jet S |
| Type | NiMH Battery |
| For Use With (Equipment) | accu-jet S and pro Pipet Controller |