Learn More
Thermo Scientific™ Accucore™ 150 C18 HPLC Columns
Use this column, with its specially designed solid core, for high-resolution separation of biomolecules.
430.00€ - 1291.32€
Specifications
For Use With | HPLC |
---|---|
Particle Size | 2.6 μm |
pH | 1 to 11 |
Pore Size | 150 Å |
Surface Area | 80 m²/g |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
12635685
|
Thermo Scientific™
16126-032130 |
2.6 μm | 2.1 mm | 30 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12645685
|
Thermo Scientific™
16126-034630 |
2.6 μm | 4.6 mm | 30 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12675685
|
Thermo Scientific™
16126-052130 |
2.6 μm | 2.1 mm | 50 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12605695
|
Thermo Scientific™
16126-102130 |
2.6 μm | 2.1 mm | 100 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12695685
|
Thermo Scientific™
16126-103030 |
2.6 μm | 3 mm | 100 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12615695
|
Thermo Scientific™
16126-104630 |
2.6 μm | 4.6 mm | 100 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12625695
|
Thermo Scientific™
16126-152130 |
2.6 μm | 2.1 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12635695
|
Thermo Scientific™
16126-153030 |
2.6 μm | 3 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12645695
|
Thermo Scientific™
16126-154630 |
2.6 μm | 4.6 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12675695
|
Thermo Scientific™
16126-014005 |
2.6 μm | 4 mm | 10 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12665695
|
Thermo Scientific™
16126-013005 |
2.6 μm | 3 mm | 10 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12655695
|
Thermo Scientific™
16126-012105 |
2.6 μm | 2.1 mm | 10 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12635715
|
Thermo Scientific™
16126-157569 |
2.6 μm | 0.075 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12645715
|
Thermo Scientific™
16126-507569 |
2.6 μm | 0.075 mm | 500 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
Offer phase characteristics designed for the separation of peptides with Thermo Scientific™ Accucore™ 150 C18 LC Columns. Based on Core Enhanced Technology, Accucore columns provide fast, high-resolution biomolecule separations without the elevated backpressures required by sub-2μm particles.
- 150Å pore size solid core particles for fast biomoleucule separations
- Superb resolution at low backpressures
- Exceptionally rugged columns
- Designed for the separation of peptides Outstanding resolution
Ordering Information:
Other custom column dimensions are available. Please call your local Customer Service for more information.
Specifications
HPLC | |
1 to 11 | |
80 m²/g | |
L1 | |
7% | |
Yes |
2.6 μm | |
150 Å | |
60°C | |
C18 | |
Reversed Phase | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.