missing translation for 'onlineSavingsMsg'
Learn More

Acemetacin Ethyl Ester, Mikromol™

Product Code. 30051113
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30051113

Brand: Mikromol MM1728.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Acemetacin Ethyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acemetacin Ethyl Ester
CAS 76812-43-6
Analyte or Component Names Acemetacin Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C23 H22 Cl N O6
Certifications/Compliance ISO 9001
Synonym Acemetacin Ethyl Ester, 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid 2-Ethoxy-2-oxoethyl Ester
InChI Formula InChI=1S/C23H22ClNO6/c1-4-30-22(27)13-31-21(26)12-18-14(2)25(20-10-9-17(29-3)11-19(18)20)23(28)15-5-7-16(24)8-6-15/h5-11H,4,12-13H2,1-3H3
SMILES CCOC(=O)COC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13
IUPAC Name (2-ethoxy-2-oxoethyl) 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
Molecular Weight (g/mol) 443.88
Quantity 25 mg
Formula Weight 443.1136 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.