missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid-d{4}, 99.5% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7050315
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1186-52-3
C2H4O2
64.08
MFCD00051051
QTBSBXVTEAMEQO-GUEYOVJQSA-N
acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
2723903
[2H]OC(=O)C([2H])([2H])[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1186-52-3
C2H4O2
64.08
MFCD00051051
QTBSBXVTEAMEQO-GUEYOVJQSA-N
acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
2723903
[2H]OC(=O)C([2H])([2H])[2H]

Acetic acid-d{4} is used as a deuterated, polar and protic solvent, in food preservatives, PH control agent, flavor enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Density 1.12 g/mL
Flash Point 40°C (104°F)
Odor Pungent, makes eyes water
Sensitivity Moisture
Quantity 5 g
Melting Point 17°C
Boiling Point 118°C
UN Number UN2789
Solubility Information Soluble in water,ethanol and ether.
IUPAC Name deuterio 2,2,2-trideuterioacetate
Formula Weight 64.08
Percent Purity 99.5%
Chemical Name or Material Acetic acid-d4, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'