missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, sodium salt, 99%, for HPLC, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4813299
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O
This item is not returnable. View return policy
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
[Na+].CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY
Melting Point 324.0°C
Flash Point >250°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CO2Na
Quantity 50 g
Merck Index 15,8709
Solubility Information Solubility in water: 500g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name sodium;acetate
Formula Weight 82.03
Percent Purity 99%
Grade HPLC
Insoluble Matter 0.005% max
Chemical Name or Material Acetic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'