missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Honeywell Burdick & Jackson™ Biosyn™

Catalog Number 16851240
95.95 EUR valid until 2024-12-31
Use promo code "23289" to get your promotional price.


Click to view available options
Packaging:
Glass Bottle
Quantity:
2.5 L
4 x 2.5 L
This item is not returnable. View return policy
41.05
Methyl Cyanide, Cyanomethane
This item is not returnable. View return policy
41.05
Methyl Cyanide, Cyanomethane

For biosynthesis and applications that require low water content. Meets UV specifications, > 99.9% purity.

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY
Melting Point -46°C
Absorbance 0.05 AU (200 nm)
Color Colorless
CAS 75-05-8
CAS Max % 100
UN Number UN1648
Solubility Information Completely Soluble (in water)
Percent Purity ≥99.9%
Grade Low Water
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1.013 hPa)
Quantity 2.5 L
Explosion Limit 3% to 16% (V)
Flash Point 6°C
Packaging Glass Bottle
Physical Form Liquid
Product Line BioSyn™
Refractive Index 1.3434 - 1.3446 (20°C)
Vapor Pressure 97.3 hPa (20°C)
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'