Learn More
Waters Corporation ACQUITY CSH C18 Method Transfer Kit
Product Code. 17103162
Encompass
Click to view available options
Particle Size:
1.7 to 3.5 μm
1.7 to 5 μm
Type:
Method Transfer Kit
Method Transfer Kit, High Res
Unit Size:
Pack of 2
Product Code. 17103162
Brand: Waters Corporation 186005535
ACQUITY Method Transfer Kits contain an ACQUITY CSH column and matching XSelect HPLC.
  • The columns contained in an ACQUITY CSH Method Transfer Kit possess the same length to particle size (L/dp) ratio and will provide the same selectivity.
Specifications
Particle Shape Spherical
Particle Size 1.7 to 3.5 μm
Pore Size 130 Å
Quantity 2/Pk.
USP Type L1
Endcapped Yes
Type Method Transfer Kit, High Res