missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation ACQUITY UPLC BEH125 SEC Guard Column

This guard column kit contains an advanced, ultra-low volume guard device and guard connection tubing designed specifically for use in UPLC SEC applications up to 6000 psi (400 bar).

Brand:  Waters Corporation 186006504

Additional Details : Weight : 0.11140kg

Product Code. 17143752

  • 1042.50€ / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket

Description

Description

  • This guard column kit contains an advanced, ultra-low volume guard device (4.6 mm ID x 30 mm) containing BEH125, SEC, 1.7 μm particles and guard connection tubing (0.005 x 1.75 in.) designed specifically for use in UPLC SEC applications up to 6000 psi (400 bar).
Specifications

Specifications

1/Ea.
30 mm
Guard Column
Protein
No
1.7 μm
125 Å
4.6 mm
Spherical
L33
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions