Learn More
Cytiva (formerly Pall Lab) AcroPak™ 20 Filters with Fluorodyne™ II Membrane and Filling Bell
AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations.
Brand: Cytiva (formerly Pall Lab) 12201
Additional Details : Weight : 0.33250kg
Description
- Efficient processing volumes up to 20 L
- Fluorodyne II (PVDF) membrane is compatible with aqueous and many organic solvents
Specifications
Hydrophilic PVDF | |
Sterile | |
Stepped Hose Barb |
0.2 μm | |
20 cm2 | |
3/Pk. |