Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Optimized for a variety of applications requiring larger per well volume processing.
Brand: Cytiva (formerly Pall Lab) 8163
Additional Details : Weight : 0.64600kg
Description
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Omega (Modified PES) | |
Non-sterile | |
5/Pk. |
3 kDa | |
Polypropylene Housing | |
1 mL |