Learn More
Acrylamide/Bis Solution, 37.5:1, (30% w/v), 2.6% C, Serva Electrophoresis™
Product Code. 11472803
Encompass_Preferred
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
Each
Product Code. 11472803
Brand: Serva Electrophoresis 1068803
Specifications
Physical Form Liquid
Composition Acrylamide/Bis Solution, 37.5:1
Conductivity <100μS/cm
Density 1.04 at 20°Cg/cm3
Color Colorless
pH 6 to 8
Quantity 1 L
Odor Odorless
Solubility Information Fully miscible
compliance-icons compliance-icons
Product Identifier
  • Acrylamide/Bis Solution, 37.5:1
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1
  • Carcinogenicity Category 1
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H372-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.