Learn More
Deutsch Neumann™ Acrylnitril-Butadien Stoppers
Brand:  Deutsch Neumann™ 1010331
Additional Details : Weight : 0.03000kg
 View more versions of this product
Product Code. 10606371
  • 2.25€ / Each
Estimated Shipment: 05-06-2024
to see stock.
Specifications
Black
35mm
38 mm
Acrylnitril-Butadien
Stopper
31 mm
1/Pk.
45 Shore A