missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan-Actin Ab-5 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry, western blotting, and immunoprecipitation experiments.
116.00€ - 805.00€
Specifications
Antigen | Actin, pan Ab-5 |
---|---|
Clone | ACTN05; same as C4 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Form | Concentrated |
Product Code | Brand | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Conjugate | Concentration | Quantity | Price | Quantity & Availability | |||
12619496
|
Epredia
MS-1295-B1 |
Biotin | 200μg/mL | 500 μL |
535.00€
500µL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12699486
|
Epredia
MS-1295-B |
Biotin | 200μg/mL | 1 mL |
805.00€
1mL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12649496
|
Epredia
MS-1295-P1 |
Unconjugated | 200μg/mL | 500 μL |
404.00€
500µL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12659496
|
Epredia
MS-1295-P1ABX |
Unconjugated | 1mg/mL | 100 μL |
448.00€
100µL |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12609496
|
Epredia
MS-1295-B0 |
Biotin | 200μg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12629496
|
Epredia
MS-1295-P |
Unconjugated | 200μg/mL | 1 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12639496
|
Epredia
MS-1295-P0 |
Unconjugated | 200μg/mL | 100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
12669496
|
Epredia
MS-1295-PABX |
Unconjugated | 1mg/mL | 200 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
Host Species: Mouse
Clone: ACTN05; same as C4
Isotype: IgG1/κ
Species Reactivity: Human, Cow, Mouse, Dog, Pig, Rabbit, Rat, Chicken, Dictyostelium discoideum and Physarum polycephalum. Others not known.
Epitope: Not determined
Immunogen: Chicken gizzard actin
Positive Control: HeLa cells
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Denatured verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Actin, pan Ab-5 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Concentrated | |
RUO | |
Bovine, Canine, Chicken, Human, Mouse, P. polycephalum, Porcine, Rabbit, Rat | |
IgG1 κ |
ACTN05; same as C4 | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Chicken gizzard actin | |
Primary |