Learn More
Atlas Material Testing™ Activated Carbon Cartridge
Click to view available options
:
Threshold: 25μM
Specifications
Specifications
| Flow Rate | 600 L/hr. |
| Temperature (Metric) Max. Operating | 35°C |
| Temperature (Metric) Operating | 35°C |
| Length (Metric) | 254 mm |