Learn More
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Catalog Number 10127510
Click to view available options
Quantity:
250 g
500 g
Packaging:
Glass bottle
7440-44-0
C
12
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
7440-44-0
C
12
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
5462310
CHEBI:27594
carbon
[C]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
TRUSTED_SUSTAINABILITY
Melting Point 3652.0°C
Color Black
pH 5.5 to 8
Loss on Drying 15% max.
Packaging Glass bottle
Quantity 500 g
Merck Index 15, 1807
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in organic solvents
Formula Weight 12
Physical Form Powder
Chemical Name or Material Activated charcoal,NORIT* CN 1, NORIT CN 1
Show More Show Less
compliance-icons
Product Identifier
  • Activated charcoal
Signal Word
  • Warning
Hazard Category
  • Self-heating substance Category 2
Hazard Statement
  • H252-Self-heating in large quantities; may catch fire.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.