Learn More
Memmert™ Active Microprocessor Control
Product Code. 15968022 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
ICO150MED
Unit Size:
Each
Product Code. 15968022
Brand: Memmert™ ICO150medK7
Only null left
Add to basket
Add to basket
Memmert™ Active Microprocessor Control ensures more rapid reaching of set humidity and very short recovery times while avoiding condensate formation.
  • Humidity supply with distilled water (from an external tank) by a self-priming pump
  • Integral bacteria block by generating hotsteam
  • Dehumidifying via sterile filter
Specifications
Description 40 - 97 % rh
For Use With (Equipment) ICO150MED
Relative Humidity 40 to 97%
Type Humidity Supply
For Use With (Application) Humidifying and dehumidifying
Includes Digital indication and auto-diagnostic system