missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford ADA
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- Substance Name: [N-(Carbamoylmethyl) iminodiacetic acid)] [N-(2-Aceta-mido)iminodiacetic acid]
- CAS Number: 26239-55-4
- Molecular Formula: C6H10N2O5
- Molecular Weight: 190.2
Specifications
Specifications
| Chemical Name or Material | ADA, [N-(Carbamoylmethyl) iminodiacetic acid) |
| CAS | 26239-55-4 |
| pH | 2.0 - 4.0 |
| Physical Form | Powder |
| Quantity | 100 g |
| Molecular Formula | C6H10N2O5 |
| Molecular Weight (g/mol) | 190.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction