Learn More
Zeiss™ Adaptation Kit
Click to view available options
For Use With (Equipment):
Axio Scope.A1 Microscope
Unit Size:
Each
Description
Set of compensation plates, to be inserted instead of the tube lens of Axio Scope.A1 tubes/photo tubes for the main observer only
- Threaded mount includes:
- Glass plate, fixed
- Glass plate, exchangeable
- Attenuation filter, loosely supplied
- Attenuation filter can be used instead of exchangeable glass plate for brightness compensation between main observer and several co-observers
Specifications
Specifications
| Type | Adaptation Kit |
| For Use With (Application) | Brightness compensation between main observer and several co-observers |
| For Use With (Equipment) | Axio Scope.A1 Microscope |
| Quantity | 1 Ea. |