Learn More
Zeiss™ Adaptation Kit
Product Code. 11825103
Click to view available options
For Use With (Equipment):
Axio Scope.A1 Microscope
Unit Size:
Each
Product Code. 11825103
Brand: Zeiss™ 4251459070000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Adaptation set for tubes designed for use with Axio Scope.A1 Microscope.
Set of compensation plates, to be inserted instead of the tube lens of Axio Scope.A1 tubes/photo tubes for the main observer only
  • Threaded mount includes:
  • Glass plate, fixed
  • Glass plate, exchangeable
  • Attenuation filter, loosely supplied
  • Attenuation filter can be used instead of exchangeable glass plate for brightness compensation between main observer and several co-observers
Specifications
Type Adaptation Kit
For Use With (Application) Brightness compensation between main observer and several co-observers
For Use With (Equipment) Axio Scope.A1 Microscope
Quantity 1 Ea.