Learn More
HUBER Adapter for Bath
Accessory for thermostats.
Brand: HUBER 6360
Additional Details : Weight : 0.05000kg
Description
- Adapter
Specifications
Thread Size: M16 x 1 female to R 1/2 male | |
Thermostat | |
Adapter |