Learn More
Thermo Scientific™ DIN Tubes Adapter for Heraeus™ Centrifuges
Centrifuge decanting adapter for holding DIN tubes.
Brand: Thermo Scientific™ 75006453
Additional Details : Weight : 0.20000kg
Description
Centrifuge decanting adapter for holding DIN tubes.
Centrifuge tube decanting aid for Heraeus™ Multifuge 3S Plus, 3S-R Plus, 3L, 3L-R benchtop centrifuge.
- Compatible with DIN tubes with decanting aid
Specifications
Adapter with Decanting Aid | |
Heraeus™ Multifuge 3S Plus, 3S-R Plus, 3L, 3L-R Benchtop Centrifuge | |
28 |
7 mL | |
Yellow |